Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 3 - 4 | 368 | 116 | |
GenBank mRNA | AB086192.2 | - | 3 - 4 | 416 | 116 | |
BX931986.1 | - | 3 - 4 | 469 | 116 | ||
CR354251.1 | - | 3 - 4 | 347 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
