Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 4 - 5 | 494 | 93 | |
GenBank mRNA | AB086192.2 | - | 4 - 5 | 542 | 93 | |
BX931986.1 | - | 4 - 5 | 595 | 93 | ||
CR354251.1 | - | 4 - 5 | 473 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
