Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 7 - 8 | 708 | 150 | |
GenBank mRNA | AB086192.2 | - | 7 - 8 | 756 | 150 | |
BX931986.1 | - | 7 - 8 | 809 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
