Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 9 - 10 | 953 | 144 | |
GenBank mRNA | AB086192.2 | - | 9 - 10 | 1001 | 144 | |
BX931986.1 | - | 9 - 10 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
