Assay Details
Gene Symbol: ART7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114504.1 | NP_001107976.1 | 2 - 3 | 830 | 77 | |
GenBank mRNA | DQ777765.1 | - | 2 - 3 | 830 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
