Assay Details
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114638.1 | NP_001108110.1 | 2 - 3 | 291 | 87 | |
XM_015295139.2 | XP_015150625.1 | 2 - 3 | 290 | 87 | ||
XM_025141583.1 | XP_024997351.1 | 2 - 3 | 298 | 87 | ||
XM_025141584.1 | XP_024997352.1 | 2 - 3 | 352 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
