Assay Details
Gene Symbol: LPAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115082.1 | NP_001108554.1 | 1 - 2 | 59 | 92 | |
XM_015280609.2 | XP_015136095.1 | 2 - 3 | 251 | 92 | ||
XM_015280610.2 | XP_015136096.1 | 2 - 3 | 350 | 92 | ||
GenBank mRNA | BX930927.2 | - | 1 - 2 | 103 | 92 | |
BX935575.1 | - | 2 - 3 | 220 | 92 | ||
EU339317.1 | - | 1 - 2 | 59 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
