Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204300.1 | NP_989631.1 | 11 - 12 | 2155 | 92 | |
XM_015283590.2 | XP_015139076.1 | 10 - 11 | 2210 | 92 | ||
XM_015283596.2 | XP_015139082.1 | 10 - 11 | 2209 | 92 | ||
XM_015283598.2 | XP_015139084.1 | 10 - 11 | 2351 | 92 | ||
XM_015283599.2 | XP_015139085.1 | 7 - 8 | 1774 | 92 | ||
XM_015283601.2 | XP_015139087.1 | 10 - 11 | 1515 | 92 | ||
XM_015283602.2 | XP_015139088.1 | 10 - 11 | 1506 | 92 | ||
XM_015283605.2 | XP_015139091.1 | 10 - 11 | 1649 | 92 | ||
XM_015283606.2 | XP_015139092.1 | 9 - 10 | 1391 | 92 | ||
XM_015283607.2 | XP_015139093.1 | 9 - 10 | 1535 | 92 | ||
XM_025148476.1 | XP_025004244.1 | 10 - 11 | 2164 | 92 | ||
XM_025148477.1 | XP_025004245.1 | 10 - 11 | 2155 | 92 | ||
XM_025148478.1 | XP_025004246.1 | 10 - 11 | 2128 | 92 | ||
XM_025148479.1 | XP_025004247.1 | 9 - 10 | 1394 | 92 | ||
GenBank mRNA | AB017437.1 | - | 10 - 11 | 1679 | 92 | |
AB035522.1 | - | 9 - 10 | 1568 | 92 | ||
AB035523.1 | - | 6 - 7 | 1213 | 92 | ||
AB057641.1 | - | 11 - 12 | 2155 | 92 | ||
CR386583.1 | - | 4 - 5 | 344 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
