Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204300.1 | NP_989631.1 | 12 - 13 | 2227 | 56 | |
XM_015283596.2 | XP_015139082.1 | 11 - 12 | 2281 | 56 | ||
XM_015283602.2 | XP_015139088.1 | 11 - 12 | 1578 | 56 | ||
XM_015283605.2 | XP_015139091.1 | 11 - 12 | 1721 | 56 | ||
XM_015283606.2 | XP_015139092.1 | 10 - 11 | 1463 | 56 | ||
XM_015283607.2 | XP_015139093.1 | 10 - 11 | 1607 | 56 | ||
GenBank mRNA | AB017437.1 | - | 11 - 12 | 1751 | 56 | |
AB035522.1 | - | 10 - 11 | 1640 | 56 | ||
AB057641.1 | - | 12 - 13 | 2227 | 56 | ||
CR386583.1 | - | 5 - 6 | 416 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
