Assay Details
Gene Symbol: MAGOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308098.1 | NP_001295027.1 | 2 - 3 | 184 | 63 | |
GenBank mRNA | AF383945.1 | - | 2 - 3 | 165 | 63 | |
BU106550.1 | - | 2 - 3 | 158 | 63 | ||
BU285565.1 | - | 2 - 3 | 189 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
