Assay Details
Gene Symbol: CBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204332.1 | NP_989663.1 | 3 - 4 | 450 | 91 | |
XM_015299477.2 | XP_015154963.1 | 4 - 5 | 591 | 91 | ||
XM_015299478.2 | XP_015154964.1 | 3 - 4 | 492 | 91 | ||
GenBank mRNA | AB005617.1 | - | 3 - 4 | 450 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
