Assay Details
Gene Symbol: TIAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288708.2 | XP_015144194.1 | 11 - 12 | 1266 | 62 | |
XM_015288709.2 | XP_015144195.1 | 11 - 12 | 1187 | 62 | ||
XM_025152001.1 | XP_025007769.1 | 11 - 12 | 1317 | 62 | ||
XM_025152002.1 | XP_025007770.1 | 8 - 9 | 720 | 62 | ||
XM_025152003.1 | XP_025007771.1 | 7 - 8 | 682 | 62 |
Gene Symbol: RGS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288909.2 | XP_015144395.1 | 3 - 4 | 352 | 62 |
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY189904.1 | - | 11 - 12 | 1050 | 62 | |
BX935380.2 | - | 7 - 8 | 709 | 62 | ||
CR388906.1 | - | 7 - 8 | 716 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015288909.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
