Assay Details
Gene Symbol: TIAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288708.2 | XP_015144194.1 | 9 - 10 | 1000 | 109 | |
XM_015288709.2 | XP_015144195.1 | 9 - 10 | 921 | 109 | ||
XM_025152001.1 | XP_025007769.1 | 9 - 10 | 1051 | 109 | ||
XM_025152002.1 | XP_025007770.1 | 6 - 7 | 454 | 109 | ||
XM_025152003.1 | XP_025007771.1 | 5 - 6 | 416 | 109 |
Gene Symbol: RGS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288909.2 | XP_015144395.1 | 1 - 2 | 86 | 109 |
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY189904.1 | - | 9 - 10 | 109 | ||
BX935380.2 | - | 5 - 6 | 109 | |||
CR388906.1 | - | 5 - 6 | 450 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015288909.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
