Assay Details
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204409.1 | NP_989740.1 | 3 - 4 | 452 | 80 | |
GenBank mRNA | AB105911.2 | - | 3 - 4 | 492 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
