Assay Details
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204456.1 | NP_989787.1 | 5 - 6 | 1259 | 90 | |
XM_025149470.1 | XP_025005238.1 | 5 - 6 | 849 | 90 | ||
XM_025149471.1 | XP_025005239.1 | 5 - 6 | 830 | 90 | ||
XM_025149472.1 | XP_025005240.1 | 4 - 5 | 697 | 90 | ||
GenBank mRNA | AJ535114.1 | - | 5 - 6 | 1259 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
