Assay Details
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204456.1 | NP_989787.1 | 6 - 7 | 1616 | 86 | |
XM_025149470.1 | XP_025005238.1 | 6 - 7 | 1206 | 86 | ||
XM_025149471.1 | XP_025005239.1 | 6 - 7 | 1187 | 86 | ||
XM_025149472.1 | XP_025005240.1 | 5 - 6 | 1054 | 86 | ||
GenBank mRNA | AJ535114.1 | - | 6 - 7 | 1616 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
