Assay Details
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205106.2 | NP_990437.2 | 3 - 4 | 329 | 71 | |
GenBank mRNA | AJ441772.1 | - | 3 - 4 | 329 | 71 | |
BX934562.1 | - | 2 - 3 | 108 | 71 | ||
CV859154.1 | - | 2 - 3 | 89 | 71 | ||
M20817.1 | - | 3 - 4 | 187 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
