Assay Details
Gene Symbol: NR1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205205.2 | NP_990536.2 | 4 - 5 | 1256 | 138 | |
XM_015281306.2 | XP_015136792.1 | 5 - 6 | 1259 | 138 | ||
XM_015281307.2 | XP_015136793.1 | 5 - 6 | 1259 | 138 | ||
GenBank mRNA | CR387346.1 | - | 1 - 2 | 310 | 138 | |
X80258.1 | - | 1186 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
