Assay Details
Gene Symbol: NR1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205205.2 | NP_990536.2 | 5 - 6 | 1445 | 107 | |
XM_015281306.2 | XP_015136792.1 | 6 - 7 | 1448 | 107 | ||
GenBank mRNA | X80258.1 | - | 1375 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
