Assay Details
Gene Symbol: NCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205265.1 | NP_990596.1 | 11 - 12 | 1596 | 69 | |
GenBank mRNA | M21791.1 | - | 2 - 3 | 278 | 69 | |
X17199.1 | - | 1596 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
