Assay Details
Gene Symbol: NCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205265.1 | NP_990596.1 | 10 - 11 | 1460 | 76 | |
GenBank mRNA | M21791.1 | - | 1 - 2 | 142 | 76 | |
X17199.1 | - | 1460 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
