Assay Details
Gene Symbol: SBDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006211.2 | NP_001006211.1 | 1 - 2 | 162 | 81 | |
GenBank mRNA | AJ720650.1 | - | 1 - 2 | 162 | 81 | |
CR352680.1 | - | 1 - 2 | 99 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
