Assay Details
Gene Symbol: SBDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006211.2 | NP_001006211.1 | 3 - 4 | 496 | 123 | |
GenBank mRNA | AJ720650.1 | - | 3 - 4 | 496 | 123 | |
CR352680.1 | - | 4 - 5 | 355 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
