Assay Details
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006297.2 | NP_001006297.2 | 2 - 3 | 211 | 112 | |
XM_015296411.2 | XP_015151897.1 | 2 - 3 | 203 | 112 | ||
GenBank mRNA | AJ720403.1 | - | 2 - 3 | 214 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
