Assay Details
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006490.2 | NP_001006490.1 | 1 - 2 | 109 | 74 | |
GenBank mRNA | AJ720567.1 | - | 1 - 2 | 110 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
