Assay Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006508.1 | NP_001006508.1 | 3 - 4 | 442 | 65 | |
XM_015289158.2 | XP_015144644.1 | 3 - 4 | 420 | 65 | ||
XM_015289159.2 | XP_015144645.1 | 3 - 4 | 467 | 65 | ||
XM_025152156.1 | XP_025007924.1 | 2 - 3 | 330 | 65 | ||
GenBank mRNA | AJ719999.1 | - | 3 - 4 | 442 | 65 | |
BX929478.2 | - | 2 - 3 | 299 | 65 | ||
CR353667.1 | - | 3 - 4 | 370 | 65 | ||
HF586485.1 | - | 2 - 3 | 313 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
