Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006573.1 | NP_001006573.1 | 4 - 5 | 468 | 77 | |
XM_015294804.2 | XP_015150290.1 | 2 - 3 | 262 | |||
XM_025155117.1 | XP_025010885.1 | 3 - 4 | 356 | 77 | ||
GenBank mRNA | AJ719414.1 | - | 4 - 5 | 468 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015294804.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
