Assay Details
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007079.1 | NP_001007080.1 | 2 - 3 | 181 | 79 | |
GenBank mRNA | AJ621249.1 | - | 2 - 3 | 181 | 79 | |
CR388898.1 | - | 2 - 3 | 502 | 79 | ||
GU119892.1 | - | 2 - 3 | 181 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
