Assay Details
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007084.1 | NP_001007085.1 | 1 - 2 | 143 | 118 | |
GenBank mRNA | AJ621252.1 | - | 1 - 2 | 143 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
