Assay Details
Gene Symbol: DAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007473.1 | NP_001007474.1 | 2 - 3 | 400 | 116 | |
GenBank mRNA | U83627.1 | - | 2 - 3 | 400 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
