Assay Details
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007871.1 | NP_001007872.1 | 4 - 5 | 549 | 102 | |
GenBank mRNA | AJ720027.1 | - | 4 - 5 | 549 | 102 | |
BX929274.1 | - | 4 - 5 | 560 | 102 | ||
BX930542.1 | - | 1 - 2 | 120 | 102 | ||
BX930558.2 | - | 1 - 2 | 100 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
