Assay Details
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204760.1 | NP_990091.1 | 4 - 5 | 903 | 72 | |
GenBank mRNA | AF082192.1 | - | 4 - 5 | 903 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
