Assay Details
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204778.1 | NP_990109.1 | 12 - 13 | 1500 | 90 | |
XM_015279900.2 | XP_015135386.1 | 13 - 14 | 1680 | 90 | ||
XM_015279904.2 | XP_015135390.1 | 13 - 14 | 1674 | 90 | ||
XM_015279909.2 | XP_015135395.1 | 13 - 14 | 1607 | 90 | ||
GenBank mRNA | AF166094.1 | - | 12 - 13 | 1500 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
