Assay Details
Gene Symbol: TSC22D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205111.1 | NP_990442.1 | 1 - 2 | 220 | 130 | |
GenBank mRNA | D49740.1 | - | 1 - 2 | 220 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
