Assay Details
Gene Symbol: POLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025354.3 | NP_001020525.2 | 3 - 4 | 377 | 101 | |
GenBank mRNA | AJ851691.1 | - | 1 - 2 | 96 | 101 | |
BX933464.2 | - | 3 - 4 | 334 | 101 | ||
CR352579.1 | - | 3 - 4 | 348 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ851691.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
