Assay Details
Gene Symbol: SOCS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127314.1 | NP_001120786.1 | 1 - 2 | 50 | 109 | |
GenBank mRNA | EF544592.1 | - | 1 - 2 | 50 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001127314.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
