Assay Details
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130739.1 | NP_001124211.1 | 7 - 8 | 930 | 69 | |
GenBank mRNA | EU850803.1 | - | 7 - 8 | 930 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
