Assay Details
Gene Symbol: ANGPTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135122.1 | NP_001128594.1 | 3 - 4 | 759 | 62 | |
GenBank mRNA | AB360605.1 | - | 3 - 4 | 759 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
