Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115081.2 | NP_001108553.2 | 1 - 2 | 219 | 72 | |
GenBank mRNA | BM490170.1 | - | 1 - 2 | 219 | 72 | |
BX930625.1 | - | 1 - 2 | 161 | 72 | ||
EU339318.1 | - | 1 - 2 | 38 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
