Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044678.1 | NP_001038143.1 | 4 | 448 | 108 | |
GenBank mRNA | BX930599.1 | - | 4 | 465 | 108 | |
CR353770.1 | - | 4 | 472 | 108 | ||
DQ358721.1 | - | 4 | 448 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
