Assay Details
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204813.1 | NP_990144.1 | 5 | 1153 | 113 | |
GenBank mRNA | AJ250362.1 | - | 5 | 1153 | 113 | |
LC389196.1 | - | 5 | 1082 | 113 | ||
X53092.1 | - | 1133 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
