Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205508.1 | NP_990839.1 | 4 | 1448 | 93 | |
GenBank mRNA | M37000.1 | - | 4 | 1448 | 93 | |
Y00659.1 | - | 4 | 1448 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
