Assay Details
Gene Symbol: RPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321597.1 | NP_001308526.1 | 2 - 3 | 98 | 120 | |
GenBank mRNA | BU225803.1 | - | 2 - 3 | 99 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
