Assay Details
Gene Symbol: MRPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039913.1 | NP_001035002.1 | 2 - 3 | 177 | 147 | |
XM_015286027.2 | XP_015141513.2 | 2 - 3 | 334 | 147 | ||
GenBank mRNA | AB194405.1 | - | 2 - 3 | 177 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001039913.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
