Assay Details
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044666.1 | NP_001038131.1 | 1 - 2 | 69 | 82 | |
GenBank mRNA | DQ358722.1 | - | 1 - 2 | 69 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
