Assay Details
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044666.1 | NP_001038131.1 | 2 - 3 | 173 | 60 | |
GenBank mRNA | BX934459.2 | - | 2 - 3 | 171 | 60 | |
CR352538.1 | - | 2 - 3 | 260 | 60 | ||
DQ358722.1 | - | 2 - 3 | 173 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
