Assay Details
Gene Symbol: OSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170344.1 | NP_001163815.1 | 2 - 3 | 1003 | 80 | |
XM_015282958.2 | XP_015138444.1 | 3 - 4 | 1291 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
