Assay Details
Gene Symbol: APC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171771.1 | NP_001165242.1 | 11 - 12 | 1495 | 90 | |
GenBank mRNA | GQ131429.1 | - | 11 - 12 | 1495 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
