Assay Details
Gene Symbol: APC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171771.1 | NP_001165242.1 | 13 - 14 | 1690 | 103 | |
GenBank mRNA | GQ131429.1 | - | 13 - 14 | 1690 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
