Assay Details
Gene Symbol: APC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171771.1 | NP_001165242.1 | 1 - 2 | 84 | 123 | |
GenBank mRNA | GQ131429.1 | - | 1 - 2 | 84 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001171771.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
